Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEIOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163560.2 | NP_001157032.1 | 9, 10 | 979 | ||
NM_152764.2 | NP_689977.2 | 9, 10 | 902 | |||
GenBank mRNA | BC029829.1 | - | 5, 6 | 504 | 5171851 | |
CD300613.1 | - | 6, 7 | 657 |