Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175900.3 | NP_787096.2 | 2 | 241 | ||
GenBank mRNA | AK093000.1 | - | 2 | 228 | ||
AY358833.1 | - | 2 | 225 | |||
BC104466.2 | - | 2 | 210 | 40032034 | ||
BC104467.2 | - | 2 | 210 | 40032037 | ||
DA427069.1 | - | 2 | 241 |