Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIMKLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173642.3 | NP_775913.2 | 5 | 995 | ||
XM_006710585.3 | XP_006710648.1 | 5 | 873 | |||
XM_017001082.1 | XP_016856571.1 | 6 | 1054 | |||
GenBank mRNA | BC039737.1 | - | 5 | 989 | 6021672 |