Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319039.1 | NP_001305968.1 | 2 | 447 | ||
NM_001319041.1 | NP_001305970.1 | 2 | 447 | |||
NM_001319042.1 | NP_001305971.1 | 2 | 188 | |||
NM_001446.4 | NP_001437.1 | 2 | 447 | |||
GenBank mRNA | AB208815.1 | - | 2 | 194 | ||
AI879148.1 | - | 2 | 490 | |||
AJ002962.1 | - | 2 | 203 | |||
AK289836.1 | - | 2 | 206 | |||
AK307272.1 | - | 2 | 207 | |||
AK311867.1 | - | 2 | 206 | |||
AL512688.1 | - | 2 | 163 | |||
BC012299.1 | - | 2 | 313 | 4707233 | ||
BI860090.1 | - | 2 | 421 | |||
BX461253.2 | - | 2 | 168 | |||
CR457057.1 | - | 2 | 127 | |||
D50373.1 | - | 2 | 179 | |||
D88648.1 | - | 2 | 178 | |||
KM111186.1 | - | 2 | 188 | |||
U51338.1 | - | 2 | 182 | |||
U81235.1 | - | 2 | 142 |