Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020949.2 | NP_066000.2 | 8 | 7912 | ||
GenBank mRNA | AK094547.1 | - | 1 | 328 |