Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207352.3 | NP_997235.3 | 7 | 1250 | ||
XM_005262935.3 | XP_005262992.1 | 7 | 1035 | |||
XM_017008037.1 | XP_016863526.1 | 6 | 1008 | |||
GenBank mRNA | AK122600.1 | - | 7 | 985 | ||
AK126473.1 | - | 8 | 919 | |||
AY422002.1 | - | 7 | 1250 | |||
BC060857.1 | - | 7 | 1116 | 30333559 | ||
BX648730.1 | - | 3 | 1156 |