Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004125.2 | NP_001004125.1 | 1 | 1873 | ||
GenBank mRNA | AY168647.2 | - | 1 | 1455 | ||
BC017510.1 | - | 1 | 1248 | |||
BC028316.1 | - | 1 | 972 | |||
BC036931.1 | - | 1 | 951 | |||
BC046919.1 | - | 1 | 1314 | |||
BC110824.1 | - | 1 | 1455 | 5169320 |