Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001479.3 | NP_001001479.1 | 2 | 1589 | ||
XM_017028794.1 | XP_016884283.1 | 3 | 2681 | |||
XM_017028795.1 | XP_016884284.1 | 3 | 2255 | |||
GenBank mRNA | BC040191.1 | - | 2 | 1597 | 4814106 | |
BC107119.2 | - | 2 | 975 | 40023535 | ||
CR456351.1 | - | 2 | 1078 |