Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPER4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198137.1 | NP_937770.1 | 7 | 966 | ||
XM_011541432.2 | XP_011539734.1 | 7 | 1073 | |||
GenBank mRNA | BC128138.1 | - | 7 | 917 | 40112485 | |
BC128139.1 | - | 7 | 966 | 40112493 | ||
BC144663.1 | - | 6 | 935 |