Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012985.2 | NP_001013003.1 | 2 | 245 | ||
NM_001206641.2 | NP_001193570.2 | 2 | 329 | |||
NM_001301733.1 | NP_001288662.1 | 1 | 616 | |||
GenBank mRNA | BC025793.1 | - | 2 | 152 | ||
BC035925.1 | - | 2 | 242 | |||
BC116455.1 | - | 2 | 245 | 4540930 | ||
BG204581.1 | - | 1 | 88 | |||
CB136291.1 | - | 2 | 298 |