Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000553.4 | NP_000544.2 | 17 | 2723 | ||
XM_011544639.2 | XP_011542941.1 | 16 | 2124 | |||
XM_011544640.1 | XP_011542942.1 | 5 | 436 | |||
GenBank mRNA | AB209652.1 | - | 3 | 166 | ||
AF091214.1 | - | 17 | 2166 | |||
AL833572.1 | - | 5 | 569 | |||
AY818673.1 | - | 16 | 1935 |