Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000061.2 | NP_000052.1 | 8 | 909 | ||
NM_001287344.1 | NP_001274273.1 | 8 | 1065 | |||
NM_001287345.1 | NP_001274274.1 | 9 | 1074 | |||
GenBank mRNA | AF153763.1 | - | 3 | 224 | ||
AK057105.1 | - | 9 | 1028 | |||
AK225577.1 | - | 8 | 871 | |||
AK309851.1 | - | 8 | 1051 | |||
AK314382.1 | - | 8 | 876 | |||
BC109079.1 | - | 8 | 841 | 40034287 | ||
BC109080.1 | - | 8 | 841 | 40034289 | ||
KF241986.1 | - | 7 | 716 | |||
X58957.1 | - | 8 | 848 |