Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014798.2 | NP_055613.1 | 12 | 5235 | ||
NR_027774.1 | 11 | 4987 | ||||
NR_027782.1 | 11 | 4850 | ||||
XM_006722201.3 | XP_006722264.1 | 10 | 5016 | |||
XM_011525524.1 | XP_011523826.1 | 12 | 5385 | |||
XM_011525525.1 | XP_011523827.1 | 11 | 4993 | |||
XM_011525526.1 | XP_011523828.1 | 13 | 5029 | |||
XM_011525528.2 | XP_011523830.1 | 7 | 3579 | |||
XM_017025451.1 | XP_016880940.1 | 12 | 5183 | |||
XM_017025452.1 | XP_016880941.1 | 11 | 4938 | |||
XM_017025453.1 | XP_016880942.1 | 13 | 5392 | |||
XM_017025454.1 | XP_016880943.1 | 11 | 5083 | |||
GenBank mRNA | AB002354.2 | - | 12 | 5188 | ||
AK074103.1 | - | 8 | 3983 | |||
AK299116.1 | - | 11 | 4935 | |||
AK299467.1 | - | 11 | 3058 | |||
BC064361.1 | - | 12 | 5199 | 5535600 | ||
X85792.1 | - | 1 | 443 |