Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTC8 SIRNA Assays ›
Gene Symbol:
TTC8
Gene Name:
tetratricopeptide repeat domain 8
Gene Aliases:
BBS8, RP51
Chromosome Location:
Chr.14: 88824153 - 88877996 on Build GRCh38
UniGene ID:
Hs.303055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s531674
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TTC8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288781.1
NP_001275710.1
12
1254
NM_001288782.1
NP_001275711.1
10
1174
NM_001288783.1
NP_001275712.1
11
1146
NM_144596.3
NP_653197.2
12
1206
NM_198309.3
NP_938051.1
11
1176
NM_198310.3
NP_938052.1
10
1086
XM_006720035.1
XP_006720098.1
10
1458
XM_006720037.2
XP_006720100.1
9
1368
XM_011536432.1
XP_011534734.1
11
1536
XM_011536433.1
XP_011534735.1
11
1536
XM_011536434.1
XP_011534736.1
10
1446
XM_011536435.2
XP_011534737.1
10
922
XM_017020977.1
XP_016876466.1
10
1458
XM_017020978.1
XP_016876467.1
9
1368
GenBank mRNA
AK093891.1
-
10
1024
AK124675.1
-
11
1056
AK315772.1
-
10
1061
AL833901.1
-
9
910
AY366523.1
-
10
1032
AY366524.1
-
9
942
AY373972.1
-
10
1032
BC001563.1
-
6
524
BC026351.1
-
11
1110
4799330
BC095433.1
-
10
1014
30368216
BX161472.1
-
9
1014
BX247959.1
-
10
1123
BX248071.1
-
2
263
BX248248.1
-
7
618
KU178849.1
-
11
1058
KU178850.1
-
10
980
KU178851.1
-
9
890
KU178852.1
-
5
500
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein transport
cilium assembly
establishment of anatomical structure orientation
sensory processing
RNA polymerase II repressing transcription factor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-qlhls:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0