Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029871.3 | NP_001025042.2 | 2 | 358 | ||
NM_001040007.2 | NP_001035096.1 | 2 | 358 | |||
XM_017027839.1 | XP_016883328.1 | 2 | 286 | |||
GenBank mRNA | AK122609.1 | - | 2 | 355 | ||
DQ355152.1 | - | 2 | 358 |