Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318876.1 | NP_001305805.1 | 3 | 324 | ||
NM_203290.3 | NP_976035.1 | 3 | 324 | |||
XM_011515000.2 | XP_011513302.1 | 3 | 265 | |||
GenBank mRNA | AF008442.1 | - | 3 | 250 | ||
AF047441.1 | - | 3 | 260 | |||
AK293561.1 | - | 3 | 250 | |||
BC008118.1 | - | 3 | 215 | |||
BC008863.2 | - | 3 | 250 | 4128104 | ||
BM828768.1 | - | 3 | 315 |