Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016239.3 | NP_057323.3 | 9 | 4461 | ||
XM_011523918.2 | XP_011522220.1 | 7 | 4815 | |||
XM_017024714.1 | XP_016880203.1 | 7 | 4815 | |||
XM_017024715.1 | XP_016880204.1 | 7 | 4824 | |||
GenBank mRNA | AF144094.1 | - | 9 | 4461 |