Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191060.1 | NP_001177989.1 | 10 | 1707 | ||
NM_001191061.1 | NP_001177990.1 | 10 | 1376 | |||
NM_024698.5 | NP_078974.1 | 10 | 1544 | |||
XM_011520369.1 | XP_011518671.1 | 10 | 1418 | |||
XM_011520370.1 | XP_011518672.1 | 10 | 1486 | |||
XM_011520371.1 | XP_011518673.1 | 10 | 1371 | |||
GenBank mRNA | AJ428202.1 | - | 9 | 1264 | ||
AK023106.1 | - | 9 | 1264 | |||
AK290481.1 | - | 10 | 1707 | |||
AL832423.1 | - | 5 | 796 | |||
BC019033.2 | - | 10 | 1342 | 4577143 | ||
BC023545.2 | - | 10 | 1502 | 4110556 | ||
BC024212.2 | - | 6 | 885 |