Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278666.1 | NP_001265595.1 | 7 | 3895 | ||
NM_001278667.1 | NP_001265596.1 | 7 | 4067 | |||
NM_001278668.1 | NP_001265597.1 | 7 | 3894 | |||
NM_016277.4 | NP_057361.3 | 7 | 4364 | |||
NM_183227.2 | NP_899050.1 | 7 | 3918 | |||
NR_103822.1 | 4 | 3569 |