Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174916.2 | NP_777576.1 | 9 | 1112 | ||
GenBank mRNA | AF525401.1 | - | 9 | 995 | ||
AK027803.1 | - | 6 | 553 | |||
AK056441.1 | - | 9 | 1062 | |||
AK292516.1 | - | 9 | 1059 | |||
AK302489.1 | - | 9 | 1139 | |||
AY061886.1 | - | 9 | 1046 | |||
BC113505.1 | - | 9 | 1124 | 8322557 | ||
BC113507.1 | - | 9 | 1124 | 8322559 |