Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2T5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004697.1 | NP_001004697.1 | 1 | 335 | ||
GenBank mRNA | BC140747.1 | - | 1 | 340 | 9021615 | |
BC140915.1 | - | 1 | 360 | 9056727 |
Gene Symbol: OR2T29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004694.2 | NP_001004694.2 | 1 | 335 |