Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR5H14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005514.1 | NP_001005514.1 | 1 | 735 | ||
GenBank mRNA | BC136984.1 | - | 1 | 795 | 9020976 |