Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001852.3 | NP_001001852.2 | 6 | 2332 | ||
GenBank mRNA | AB114795.1 | - | 6 | 2314 | ||
BC017083.2 | - | 1 | 1089 | |||
BC052239.1 | - | 6 | 1913 | |||
BC064477.1 | - | 2 | 1305 | |||
BC141855.1 | - | 6 | 1944 | 8860375 |