Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYG11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004339.2 | NP_001004339.2 | 12, 13 | 2382 | ||
NM_001307931.1 | NP_001294860.1 | 11, 12 | 1630 | |||
XM_011541480.2 | XP_011539782.1 | 11, 12 | 2112 | |||
XM_011541481.2 | XP_011539783.1 | 11, 12 | 1980 | |||
XM_011541482.2 | XP_011539784.1 | 11, 12 | 1962 | |||
XM_011541483.2 | XP_011539785.1 | 10, 11 | 1917 | |||
GenBank mRNA | AY271826.1 | - | 11, 12 | 1621 |