Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000402.4 | NP_000393.4 | 2 | 319 | ||
NM_001042351.2 | NP_001035810.1 | 2 | 208 | |||
GenBank mRNA | AK292304.1 | - | 2 | 145 | ||
AK302341.1 | - | 2 | 146 | |||
AL560686.3 | - | 2 | 194 | |||
BC000337.2 | - | 2 | 134 | 2822640 | ||
DC394789.1 | - | 2 | 208 | |||
M21248.1 | - | 1 | 95 | |||
M27940.1 | - | 2 | 318 | |||
S58359.1 | - | 2 | 234 | |||
X03674.1 | - | 2 | 551 |