Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB106B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040704.1 | NP_001035794.1 | 2 | 112 |
Gene Symbol: DEFB106A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152251.3 | NP_689464.1 | 2 | 112 | ||
GenBank mRNA | AB089181.1 | - | 2 | 107 | ||
AF529417.1 | - | 2 | 107 | |||
AF540978.1 | - | 2 | 107 | |||
AY122466.1 | - | 2 | 139 | |||
BC100844.2 | - | 2 | 112 | 40002124 | ||
BC100845.2 | - | 2 | 112 | 40002125 | ||
BC100846.2 | - | 2 | 112 | 40002126 | ||
BC100847.2 | - | 2 | 112 | 40002135 | ||
BC110062.1 | - | 2 | 112 | 40002127 | ||
BC111688.1 | - | 2 | 112 | 40002128 |