Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145101.1 | NP_001138573.1 | 3 | 718 | ||
XM_011545212.2 | XP_011543514.1 | 3 | 1977 | |||
XM_017018127.1 | XP_016873616.1 | 2 | 2417 | |||
XM_017018128.1 | XP_016873617.1 | 3 | 674 | |||
GenBank mRNA | BC157853.1 | - | 2 | 623 | 9057072 | |
BC157854.1 | - | 2 | 623 | 9057073 |