Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005248333.4 | XP_005248390.1 | 1 | 514 | ||
XM_006714486.3 | XP_006714549.1 | 1 | 514 | |||
XM_011514074.2 | XP_011512376.1 | 1 | 514 | |||
XM_011514075.2 | XP_011512377.1 | 1 | 514 |