Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM21A SIRNA Assays ›
Gene Symbol:
FAM21A
FAM21C
Gene Name:
family with sequence similarity 21 member A
family with sequence similarity 21 member C
Gene Aliases:
FAM21B, bA56A21.1, bA98I6.1
FAM21A, VPEF
Chromosome Location:
Chr.10: 50067888 - 50133509 on Build GRCh38
Chromosome Location:
Chr.10: 45727170 - 45792964 on Build GRCh38
UniGene ID:
Hs.365286
Hs.365286
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s57671
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM21A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005751.2
NP_001005751.1
29, 30
3837
NM_001291398.1
NP_001278327.1
28, 29
3774
XM_005269806.3
XP_005269863.2
28, 29
3713
XM_005269807.3
XP_005269864.2
28, 29
3698
XM_005269808.3
XP_005269865.2
27, 28
3599
XM_005269810.3
XP_005269867.2
11, 12
1930
XM_006717831.3
XP_006717894.2
28, 29
3632
XM_006717832.3
XP_006717895.2
26, 27
3497
XM_011539786.2
XP_011538088.1
26, 27
3386
XM_017016227.1
XP_016871716.1
28, 29
3662
XM_017016228.1
XP_016871717.1
27, 28
3650
XM_017016229.1
XP_016871718.1
27, 28
3635
XM_017016230.1
XP_016871719.1
28, 29
3650
XM_017016231.1
XP_016871720.1
27, 28
3569
XM_017016232.1
XP_016871721.1
26, 27
3563
XM_017016233.1
XP_016871722.1
26, 27
3360
XM_017016234.1
XP_016871723.1
28, 29
3720
XM_017016235.1
XP_016871724.1
20, 21
2736
GenBank mRNA
AK125147.1
-
1, 2
2191
AK304116.1
-
29, 30
3773
AL080183.1
-
5, 6
1169
BC065500.1
-
1, 2
75
BC075815.1
-
25, 26
3339
BC082258.1
-
29, 30
3764
6278433
BC144479.1
-
28, 29
3803
9053006
BQ060891.1
-
1, 2
597
Gene Symbol: FAM21C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001169106.1
NP_001162577.1
27, 28
3651
NM_001169107.1
NP_001162578.1
26, 27
3549
NM_015262.2
NP_056077.2
28, 29
3774
XM_011539566.1
XP_011537868.1
29, 30
3804
XM_011539567.1
XP_011537869.1
28, 29
3732
XM_011539568.1
XP_011537870.1
28, 29
3717
XM_011539569.1
XP_011537871.1
28, 29
3651
XM_011539570.2
XP_011537872.1
28, 29
3693
XM_011539571.1
XP_011537873.1
26, 27
3405
XM_011539573.2
XP_011537875.1
11, 12
1930
XM_017016014.1
XP_016871503.1
28, 29
3681
XM_017016015.1
XP_016871504.1
27, 28
3669
XM_017016016.1
XP_016871505.1
27, 28
3654
XM_017016017.1
XP_016871506.1
27, 28
3645
XM_017016018.1
XP_016871507.1
27, 28
3588
XM_017016019.1
XP_016871508.1
27, 28
3579
XM_017016020.1
XP_016871509.1
26, 27
3546
XM_017016021.1
XP_016871510.1
29, 30
3732
XM_017016022.1
XP_016871511.1
28, 29
3715
XM_017016023.1
XP_016871512.1
26, 27
3464
XM_017016024.1
XP_016871513.1
17, 18
2546
XM_017016025.1
XP_016871514.1
18, 19
2617
GenBank mRNA
AB011164.1
-
28, 29
3755
AK001545.1
-
11, 12
1998
AK291430.1
-
28, 29
3762
AK293929.1
-
24, 25
3288
AK303048.1
-
26, 27
3513
AL050279.1
-
2, 3
1509
BC006456.1
-
2, 3
702
BC039561.1
-
1, 2
199
BC150611.1
-
27, 28
3616
9056981
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_199207, XM_006237147, XM_006237149, AY257251.This siRNA also targets the following transcripts in Mouse: NM_026585, BC040393, AK168388, AK122320, AK145716, and others.
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein transport
retrograde transport, endosome to Golgi
regulation of substrate adhesion-dependent cell spreading
retrograde transport, endosome to plasma membrane
negative regulation of barbed-end actin filament capping
lipid binding
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
phosphatidylinositol-5-phosphate binding
phosphatidylinositol-3-phosphate binding
phosphatidylinositol-3,4-bisphosphate binding
phosphatidylinositol-4-phosphate binding
phosphatidylinositol-3,5-bisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-kd7lp:80/10.239.160.160:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0