Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135167.1 | 11 | 1174 | |||
XM_005268826.1 | XP_005268883.1 | 11 | 1544 | |||
GenBank mRNA | BC070315.1 | - | 11 | 1116 | 4101267 | |
X79536.1 | - | 11 | 1082 |