Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CT45A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291527.1 | NP_001278456.1 | 2 | 246 | ||
NM_001291528.1 | NP_001278457.1 | 2 | 229 | |||
NM_001291529.1 | NP_001278458.1 | 2 | 167 | |||
NM_001291530.1 | NP_001278459.1 | 2 | 184 | |||
XM_011531238.2 | XP_011529540.1 | 2 | 180 | |||
XM_011531239.1 | XP_011529541.1 | 2 | 161 | |||
GenBank mRNA | AK292207.1 | - | 2 | 163 | ||
AY743713.1 | - | 2 | 322 | |||
BC070138.1 | - | 2 | 167 | 4685945 | ||
BC144429.1 | - | 2 | 221 | 9052950 | ||
BC144430.1 | - | 2 | 238 | 9052951 | ||
BC144527.1 | - | 2 | 244 | 9053054 | ||
BC144528.1 | - | 2 | 229 | 9053055 |
Gene Symbol: CT45A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017417.2 | NP_001017417.1 | 2 | 321 | ||
XM_005278141.3 | XP_005278198.1 | 2 | 170 | |||
XM_011531352.1 | XP_011529654.1 | 1 | 91 | |||
GenBank mRNA | AY743709.1 | - | 2 | 322 | ||
BC140913.1 | - | 2 | 221 | 9056725 |
Gene Symbol: CT45A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152582.6 | NP_689795.4 | 2 | 322 | ||
XM_006724783.2 | XP_006724846.1 | 2 | 249 | |||
XM_011531389.2 | XP_011529691.1 | 2 | 217 | |||
GenBank mRNA | AY743710.1 | - | 2 | 322 |
Gene Symbol: CT45A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017435.1 | NP_001017435.1 | 2 | 322 | ||
XM_006724758.3 | XP_006724821.1 | 2 | 239 | |||
XM_011531340.2 | XP_011529642.1 | 2 | 198 | |||
GenBank mRNA | AK098689.1 | - | 2 | 167 | ||
AY743711.1 | - | 2 | 322 |
Gene Symbol: CT45A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007551.4 | NP_001007552.2 | 2 | 322 | ||
NM_001172288.2 | NP_001165759.2 | 2 | 339 | |||
XM_011531341.2 | XP_011529643.1 | 2 | 204 | |||
GenBank mRNA | BC028711.2 | - | 2 | 322 | 4828274 | |
BC140735.1 | - | 2 | 229 | 9021603 | ||
BC140926.1 | - | 2 | 229 | 9056738 | ||
BC144437.1 | - | 2 | 229 | 9052959 |
Gene Symbol: CT45A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017438.2 | NP_001017438.2 | 2 | 322 | ||
XM_005262425.4 | XP_005262482.2 | 2 | 242 | |||
XM_011531351.1 | XP_011529653.1 | 2 | 173 | |||
GenBank mRNA | AY743714.1 | - | 2 | 322 |
Gene Symbol: CT45A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291543.1 | NP_001278472.1 | 2 | 138 | ||
XM_003960082.4 | XP_003960131.1 | 2 | 225 | |||
XM_011531229.2 | XP_011529531.1 | 2 | 204 | |||
GenBank mRNA | BU540102.1 | - | 2 | 141 |
Gene Symbol: CT45A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291535.1 | NP_001278464.1 | 2 | 322 | ||
XM_006724799.2 | XP_006724862.1 | 2 | 249 | |||
XM_011531241.2 | XP_011529543.1 | 2 | 217 | |||
GenBank mRNA | BP371111.1 | - | 2 | 167 |
Gene Symbol: CT45A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291540.2 | NP_001278469.1 | 2 | 322 | ||
NM_001321271.1 | NP_001308200.1 | 2 | 167 | |||
XM_006724796.2 | XP_006724859.1 | 2 | 249 |