Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003334.3 | NP_003325.2 | 24, 25 | 3156 | ||
NM_153280.2 | NP_695012.1 | 24, 25 | 3084 | |||
XM_005272649.1 | XP_005272706.1 | 25, 26 | 3534 | |||
XM_011543954.2 | XP_011542256.1 | 25, 26 | 3176 | |||
XM_017029777.1 | XP_016885266.1 | 24, 25 | 4251 | |||
XM_017029778.1 | XP_016885267.1 | 25, 26 | 5425 | |||
XM_017029779.1 | XP_016885268.1 | 24, 25 | 5933 | |||
XM_017029780.1 | XP_016885269.1 | 24, 25 | 3041 | |||
XM_017029781.1 | XP_016885270.1 | 24, 25 | 3089 | |||
GenBank mRNA | AK097343.1 | - | 8, 9 | 2056 | ||
AK293161.1 | - | 13, 14 | 1748 | |||
AK296866.1 | - | 14, 15 | 1774 | |||
BC013041.2 | - | 24, 25 | 3023 | 3543021 | ||
BC020261.1 | - | 21, 22 | 2718 | |||
M58028.1 | - | 24, 25 | 3062 | |||
X52897.1 | - | 17, 18 | 2228 | |||
X56976.1 | - | 24, 25 | 2965 |