Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026790.1 | 1 | 1178 | |||
GenBank mRNA | AK054919.1 | - | 1 | 1036 |
Gene Symbol: LOC730020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093276.1 | - | 1 | 1021 | ||
AK098704.1 | - | 1 | 530 |