Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRB10 SIRNA Assays ›
Gene Symbol:
GRB10
Gene Name:
growth factor receptor bound protein 10
Gene Aliases:
GRB-IR, Grb-10, IRBP, MEG1, RSS
Chromosome Location:
Chr.7: 50590063 - 50793462 on Build GRCh38
UniGene ID:
Hs.164060
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s6127
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRB10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001549.2
NP_001001549.1
13
1405
NM_001001550.2
NP_001001550.1
14
1623
NM_001001555.2
NP_001001555.1
15
1870
NM_005311.4
NP_005302.3
14
1543
XM_011515304.1
XP_011513606.1
17
1982
XM_011515312.2
XP_011513614.1
16
2270
XM_011515316.1
XP_011513618.1
15
1815
XM_011515323.2
XP_011513625.1
12
3319
XM_017012029.1
XP_016867518.1
16
2202
XM_017012030.1
XP_016867519.1
15
2136
XM_017012031.1
XP_016867520.1
15
2067
XM_017012032.1
XP_016867521.1
21
2579
XM_017012033.1
XP_016867522.1
18
2117
XM_017012034.1
XP_016867523.1
16
1718
XM_017012035.1
XP_016867524.1
22
3628
XM_017012036.1
XP_016867525.1
23
3413
XM_017012037.1
XP_016867526.1
23
3250
XM_017012038.1
XP_016867527.1
22
3200
XM_017012039.1
XP_016867528.1
19
2447
XM_017012040.1
XP_016867529.1
19
2983
XM_017012041.1
XP_016867530.1
21
3104
XM_017012042.1
XP_016867531.1
17
2024
XM_017012043.1
XP_016867532.1
18
2214
XM_017012044.1
XP_016867533.1
14
1813
XM_017012045.1
XP_016867534.1
14
1477
XM_017012046.1
XP_016867535.1
18
2829
XM_017012047.1
XP_016867536.1
23
3812
XM_017012048.1
XP_016867537.1
22
3696
XM_017012049.1
XP_016867538.1
23
3318
XM_017012050.1
XP_016867539.1
22
3268
XM_017012051.1
XP_016867540.1
21
3172
XM_017012052.1
XP_016867541.1
21
3512
XM_017012053.1
XP_016867542.1
22
3134
XM_017012054.1
XP_016867543.1
21
3084
XM_017012055.1
XP_016867544.1
18
2867
XM_017012056.1
XP_016867545.1
17
3699
XM_017012057.1
XP_016867546.1
20
2988
XM_017012058.1
XP_016867547.1
16
1859
XM_017012059.1
XP_016867548.1
16
1981
XM_017012060.1
XP_016867549.1
24
3945
XM_017012061.1
XP_016867550.1
15
1811
XM_017012062.1
XP_016867551.1
14
1572
XM_017012063.1
XP_016867552.1
22
3677
XM_017012064.1
XP_016867553.1
21
3561
XM_017012065.1
XP_016867554.1
19
3013
XM_017012066.1
XP_016867555.1
17
3372
XM_017012067.1
XP_016867556.1
17
1875
XM_017012068.1
XP_016867557.1
17
2741
GenBank mRNA
AB000731.1
-
14
1518
AF000017.1
-
14
1512
AF001534.1
-
14
1543
AJ271366.1
-
17
1982
AK289511.1
-
16
1837
BC024285.1
-
15
1647
4590330
BX649028.1
-
14
1603
D86962.1
-
16
2275
U34355.1
-
13
1405
U66065.1
-
14
1623
U69276.1
-
14
1625
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of Wnt signaling pathway
positive regulation of vascular endothelial growth factor receptor signaling pathway
negative regulation of phosphorylation
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0