Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGTA1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003191.1 | 4 | 763 | |||
NR_045211.1 | 3 | 494 | ||||
GenBank mRNA | AF305838.1 | - | 2 | 107 | ||
AF378121.1 | - | LIT2093 alpha1,3GTP1 |
4 | 493 | ||
AF378122.1 | - | LIT2094 alpha1,3GTP2 |
4 | 493 | ||
AF378123.1 | - | LIT2095 alpha1,3GTP3 |
4 | 493 | ||
BC045756.1 | - | 3 | 334 | |||
DT220151.1 | - | 3 | 286 |