Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009713.4 | NP_033843.2 | 9 | 2524 | ||
GenBank mRNA | AK004540.1 | - | 8 | 1743 | ||
AK029854.1 | - | 7 | 1785 | |||
AK132501.1 | - | 8 | 1755 | |||
AK165921.1 | - | 8 | 1711 | |||
BC011284.1 | - | 8 | 1706 | 3669438 | ||
BC098075.1 | - | 9 | 2536 | 30734943 | ||
X73230.1 | - | 8 | 1957 |