Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007517.4 | NP_031543.2 | 3 | 551 | ||
GenBank mRNA | AK144497.1 | - | 1 | 503 | ||
AK154707.1 | - | 3 | 420 | |||
AK158035.1 | - | 3 | 342 | |||
AK167335.1 | - | 3 | 341 | |||
AK171011.1 | - | 3 | 354 | |||
AK171184.1 | - | 3 | 346 | |||
AK171202.1 | - | 3 | 360 | |||
AK171249.1 | - | 3 | 340 | |||
AK171541.1 | - | 3 | 341 | |||
AK171621.1 | - | 3 | 333 | |||
BC016485.1 | - | 3 | 344 | 3962982 | ||
U41736.1 | - | 3 | 349 |