Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021272.3 | NP_067247.1 | 2, 3 | 336 | ||
GenBank mRNA | AJ870970.1 | - | 2, 3 | 336 | ||
AK002955.1 | - | 2, 3 | 337 | |||
AK021271.1 | - | 2, 3 | 341 | |||
BC055280.1 | - | 2, 3 | 319 | 5710017 | ||
BC057090.1 | - | 2, 3 | 322 | 5700428 | ||
CT010401.1 | - | 2, 3 | 250 | |||
S69799.1 | - | 2, 3 | 319 |