Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009755.3 | NP_033885.2 | 1 | 366 | ||
GenBank mRNA | AK154239.1 | - | 1 | 368 | ||
AK158218.1 | - | 1 | 359 | |||
AK220295.1 | - | 1 | 340 | |||
BC053923.1 | - | 1 | 255 | |||
BC066062.1 | - | 1 | 294 | 6849066 | ||
L24755.1 | - | 1 | 144 |