Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Birc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007566.3 | NP_031592.3 | 26 | 5602 | ||
GenBank mRNA | AK134254.1 | - | 11 | 1510 | ||
Y17267.1 | - | 26 | 5603 |