Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109764.1 | NP_001103234.1 | 10 | 1505 | ||
NM_009819.2 | NP_033949.2 | 9 | 1403 | |||
NM_145732.2 | NP_663785.2 | 10 | 1505 | |||
GenBank mRNA | AK158916.1 | - | 10 | 1538 | ||
BC051995.1 | - | 9 | 1284 | |||
BC079648.1 | - | 10 | 1422 | 6850068 | ||
D25281.1 | - | 10 | 1432 | |||
D25282.1 | - | 9 | 1332 |