Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307936.1 | NP_001294865.1 | 13 | 1516 | ||
NM_018976.4 | NP_061849.2 | 14 | 1632 | |||
GenBank mRNA | AB037803.2 | - | 13 | 1192 | ||
AF259799.1 | - | 13 | 1212 | |||
AF298897.1 | - | 14 | 1543 | |||
AJ344099.1 | - | 14 | 1544 | |||
AK001700.1 | - | 13 | 1425 | |||
AK172784.1 | - | 7 | 639 | |||
AK291644.1 | - | 14 | 1336 | |||
AK293858.1 | - | 7 | 747 | |||
BC040342.1 | - | 14 | 1534 | 3874551 | ||
CR457267.1 | - | 10 | 892 |