Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009840.3 | NP_033970.3 | 13 | 1383 | ||
GenBank mRNA | AK010882.1 | - | 13 | 1384 | ||
AK014005.1 | - | 6 | 449 | |||
AK075711.1 | - | 13 | 1364 | |||
AK075880.1 | - | 13 | 1372 | |||
AK145661.1 | - | 13 | 1367 | |||
AK145754.1 | - | 13 | 1366 | |||
AK145918.1 | - | 13 | 1385 | |||
AK147038.1 | - | 13 | 1386 | |||
AK159969.1 | - | 13 | 1374 | |||
AK168465.1 | - | 13 | 1356 | |||
AK172867.1 | - | 13 | 1354 | |||
BC009007.1 | - | 13 | 1341 | 3582639 | ||
Z37164.1 | - | 13 | 1387 |