Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009969.4 | NP_034099.2 | 2 | 455 | ||
GenBank mRNA | AK053196.1 | - | 2 | 455 | ||
BC116878.1 | - | 2 | 189 | 40126197 | ||
BC116880.1 | - | 2 | 189 | 40126199 | ||
EU366957.1 | - | 2 | 166 | |||
M11848.1 | - | 2 | 124 | |||
X02333.1 | - | 2 | 339 | |||
X03019.1 | - | 2 | 198 | |||
X03221.1 | - | 2 | 197 | |||
X05906.1 | - | 2 | 97 |