Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198866.1 | NP_001185795.1 | 2 | 277 | ||
NM_001198867.1 | NP_001185796.1 | 2 | 277 | |||
NM_007835.2 | NP_031861.2 | 2 | 523 | |||
GenBank mRNA | AK157867.1 | - | 2 | 524 | ||
AK162846.1 | - | 2 | 527 | |||
AK171376.1 | - | 2 | 264 | |||
AK172261.1 | - | 2 | 251 | |||
AK194220.1 | - | 1 | 146 | |||
AK215255.1 | - | 1 | 105 | |||
BC066061.1 | - | 2 | 250 | 6849060 | ||
BY198890.1 | - | 2 | 281 | |||
BY235563.1 | - | 2 | 251 | |||
CF162175.1 | - | 2 | 234 | |||
U60312.1 | - | 2 | 258 |