Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145885.1 | NP_001139357.1 | 15 | 1176 | ||
NM_010029.2 | NP_034159.1 | 14 | 1098 | |||
GenBank mRNA | AK014844.1 | - | 14 | 1089 | ||
AK133366.1 | - | 15 | 1169 | |||
BC137601.1 | - | 14 | 1152 | 8860620 | ||
BC144760.1 | - | 14 | 1152 | 9053308 | ||
D14859.1 | - | 11 | 871 | |||
JQ923486.1 | - | 14 | 1126 |