Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007852.1 | NP_031878.1 | 1 | 70 |
Gene Symbol: Defa24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024225.2 | NP_001019396.1 | 1 | 113 | ||
GenBank mRNA | AY746429.1 | - | 1 | 70 |
Gene Symbol: Defa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | S73391.1 | - | 84 |