Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fadd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010175.5 | NP_034305.1 | 2 | 460 | ||
GenBank mRNA | AK007648.1 | - | 2 | 487 | ||
AK031396.1 | - | 2 | 461 | |||
AK084808.1 | - | 2 | 461 | |||
AK169798.1 | - | 2 | 488 | |||
AK170927.1 | - | 2 | 487 | |||
BC004584.1 | - | 2 | 450 | 3587331 | ||
BC021400.1 | - | 2 | 457 | 4190229 | ||
U43184.1 | - | 2 | 413 | |||
U50406.1 | - | 2 | 425 |