Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160239.2 | NP_001153711.1 | 2, 3 | 186 | ||
NM_001190436.1 | NP_001177365.1 | 1, 2 | 351 | |||
NM_007990.3 | NP_032016.1 | 2, 3 | 189 | |||
GenBank mRNA | AK002355.1 | - | 2, 3 | 127 | ||
AK008466.1 | - | 1, 2 | 96 | |||
AK132752.1 | - | 2, 3 | 132 | |||
AK146359.1 | - | 2, 3 | 125 | |||
AK151211.1 | - | 2, 3 | 127 | |||
AK151465.1 | - | 2, 3 | 127 | |||
AK152738.1 | - | 2, 3 | 127 | |||
AK168348.1 | - | 2, 3 | 127 | |||
AV474170.1 | - | 2, 3 | 189 | |||
BC058691.1 | - | 2, 3 | 106 | 6814998 | ||
BC062873.1 | - | 2, 3 | 118 | 5683472 | ||
BC081463.1 | - | 2, 3 | 106 | 6408436 | ||
D26610.1 | - | 2, 3 | 126 | |||
X65922.1 | - | 2, 3 | 114 |