Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010308.3 | NP_034438.1 | 7 | 1273 | ||
GenBank mRNA | AK035907.1 | - | 7 | 1274 | ||
AK047197.1 | - | 7 | 1277 | |||
BC051989.1 | - | 7 | 1100 | 5686124 | ||
M36777.1 | - | 7 | 796 |